Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
$1083.00 - $5809.07
Specifications
Material | Polypropylene |
---|---|
RNase Free | Yes |
Product Line | Nunc™ |
Dnase Free | Yes |
Catalog Number | Mfr. No. | Capacity (Metric) | Thread Style | Length (Metric) | Description | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Thread Style | Length (Metric) | Description | Price | Quantity | ||
12-570-216
|
Thermo Scientific™
374080 |
0.5 mL | Internal | 35 mm | Without writing surface |
Case of 960 Each for $3,321.59
|
|
||
12-570-217
|
Thermo Scientific™
374081 |
1 mL | Internal | 48.5 mm | With writing surface |
Case of 960 Each for $4,250.75
|
|
||
12-565-164N
|
Thermo Scientific™
375353 |
1 mL | External | 41 mm | With writing surface |
Pack of 500 Each for $1,239.45
Case of 4 Pack for $4,957.80
|
|
||
12-565-297
|
Thermo Scientific™
347597 |
1 mL | External | 41 mm | Without writing surface |
Pack of 500 Each for $1,362.02
Case of 4 Pack for $5,448.08
|
|
||
12-565-168N
|
Thermo Scientific™
377224 |
1 mL | Internal | 42 mm | With writing surface, Anti-rotational base |
Pack of 500 Each for $1,699.27
Case of 4 Pack for $6,797.08
|
|
||
12-565-169N
|
Thermo Scientific™
366656 |
1 mL | Internal | 42 mm | With writing surface |
Pack of 500 Each for $1,699.27
Case of 4 Pack for $6,797.08
|
|
||
12-565-163N
|
Thermo Scientific™
375418 |
1.8 mL | External | 48 mm | With writing surface |
Pack of 450 Each for $1,083.00
Case of 4 Pack for $4,332.00
|
|
||
12-565-298
|
Thermo Scientific™
347627 |
1.8 mL | External | 48 mm | Without writing area |
Pack of 450 Each for $1,259.50
Case of 4 Pack for $5,038.00
|
|
||
12-565-167N
|
Thermo Scientific™
377267 |
1.8 mL | Internal | 49 mm | With writing surface |
Pack of 450 Each for $1,476.20
Case of 4 Pack for $5,904.80
|
|
||
12-565-171N
|
Thermo Scientific™
368632 |
1.8 mL | Internal | 49 mm | With writing surface and screw cap |
Pack of 450 Each for $1,565.67
Case of 4 Pack for $6,262.68
|
|
||
12-565-170N
|
Thermo Scientific™
363401 |
1.8 mL | Internal | 48 mm | With writing surface and screw cap w/gasket |
Pack of 500 Each for $1,712.13
Case of 4 Pack for $6,848.52
|
|
||
12-566-97
|
Thermo Scientific™
351934 |
1.8 mL | Internal | 48 mm | With writing surface, no caps |
Case of 2500 Each for $5,809.07
|
|
||
12-565-162N
|
Thermo Scientific™
379189 |
3.6 mL | Internal | 72 mm | With writing surface |
Pack of 400 Each for $1,816.77
Case of 4 Pack for $7,267.08
|
|
||
12-565-172N
|
Thermo Scientific™
366524 |
3.6 mL | Internal | 70 mm | With writing surface |
Pack of 400 Each for $1,816.77
Case of 4 Pack for $7,267.08
|
|
||
12-566-98
|
Thermo Scientific™
367997 |
3.6 mL | Internal | 70 mm | With writing surface |
Case of 1500 Each for $4,274.87
|
|
||
12-565-291
|
Thermo Scientific™
337516 |
4.5 mL | External | 91 mm | With writing surface |
Pack of 300 Each for $1,321.41
Case of 4 Pack for $5,285.64
|
|
||
12-565-299
|
Thermo Scientific™
347643 |
4.5 mL | External | 91 mm | Without writing surface |
Pack of 300 Each for $1,326.16
Case of 4 Pack for $5,304.64
|
|
||
12-565-161N
|
Thermo Scientific™
379146 |
4.5 mL | Internal | 92 mm | With writing surface, Conical bottom |
Pack of 300 Each for $1,601.97
Case of 4 Pack for $6,407.88
|
|
||
12-565-173N
|
Thermo Scientific™
363452 |
4.5 mL | Internal | 92 mm | With writing surface, round bottom |
Pack of 300 Each for $1,697.03
Case of 4 Pack for $6,788.12
|
|
||
Description
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
Features:- Non-cytotoxic, non-mutagenic raw materials and pyrogen-free IVD compatible tubes for added sample security
- Certified SAL of 10-6 for the highest sterility assurance available
- Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
- Bulk, uncapped, non-sterile versions available for less demanding applications
Specifications
Polypropylene | |
Yes | |
Nunc™ | |
Yes |