Corning™ Polypropylene Centrifuge Tubes, Sterile
Manufactured from clear polypropylene for superior chemical resistance. Corning™ Polypropylene Centrifuge Tubes are an ideal solution for centrifuge needs.
$96.66 - $1306.79
Catalog Number | Mfr. No. | Capacity (Metric) | Max. RCF | Packaging | Closure Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Max. RCF | Packaging | Closure Type | Price | Quantity | ||
07-201-330
|
Corning™
431470 |
15 mL | 12,000 x g | No rack | Not Included |
Case of 500 Each for $536.74
|
|
||
05-538-53F
|
Corning™
430766 |
15 mL | 12,000 x g | No rack | Plug |
Case of 500 Each for $807.00
|
|
||
05-538-53D
|
Corning™
430052 |
15 mL | 12,000 x g | With rack | Plug |
Case of 500 Each for $916.37
|
|
||
07-201-332
|
Corning™
431472 |
50 mL | 12,000 x g | No rack | Not Included |
Case of 500 Each for $675.65
|
|
||
05-538-55
|
Corning™
430290 |
50 mL | 15,500 x g | With rack | Plug |
Case of 500 Each for $1,245.12
|
|
||
05-538-55A
|
Corning™
430291 |
50 mL | 15,500 x g | No rack | Plug |
Case of 500 Each for $1,097.00
|
|
||
05-538-68
|
Corning™
430921 |
50 mL | 3000 x g | No rack | CentriStar |
Case of 500 Each for $1,306.79
|
|
||
05-538-67
|
Corning™
430897 |
50 mL | 3000 x g | No rack | Plug |
Case of 500 Each for $1,285.60
|
|
||
05-538-53
|
Corning™
430776 |
250 mL | 6000 x g | No rack | Plug |
Case of 102 Each for $804.22
|
|
||
07-200-621
|
Corning™
431123 |
500 mL | 6000 x g | No rack | Plug |
Pack of 6 Each for $96.66
Case of 6 Pack for $579.96
|
|
||
Description
Manufactured from clear polypropylene for superior chemical resistance. Corning™ Polypropylene Centrifuge Tubes are an ideal solution for centrifuge needs.
- Corning centrifuge tubes feature black printed graduations and a large white marking spot.
- Available with your choice of cap styles: the advanced Corning CentriStarTM cap or the original plug seal cap.
- Sterile.
- Nonpyrogenic.
- 95 kPA (14 psi) pressure tested – plug seal cap only.
PP = polypropylene, PET = polyethylene terephthalate, RCF = relative centrifugal force (x g).
Specifications
Polypropylene | |
Sterile | |
Yes |