missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium diphosphate, 98%, extra pure, Thermo Scientific Chemicals
$63.55 - $126.24
Chemical Identifiers
CAS | 7320-34-5 |
---|---|
Molecular Formula | K4O7P2 |
Molecular Weight (g/mol) | 330.33 |
MDL Number | MFCD00011393 |
InChI Key | RYCLIXPGLDDLTM-UHFFFAOYSA-J |
Synonym | potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate |
PubChem CID | 23740 |
IUPAC Name | tetrapotassium;phosphonato phosphate |
SMILES | [K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC269230250
|
Thermo Scientific Chemicals
269230250 |
25 g | Glass Bottle |
Each for $63.55
|
|
||||
AC269230010
|
Thermo Scientific Chemicals
269230010 |
1 kg | Plastic bottle |
Each for $126.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7320-34-5 | |
330.33 | |
RYCLIXPGLDDLTM-UHFFFAOYSA-J | |
23740 | |
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O |
K4O7P2 | |
MFCD00011393 | |
potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate | |
tetrapotassium;phosphonato phosphate |
Specifications
1090°C | |
White | |
25 g | |
K4O7P2 | |
MFCD00011393 | |
potassium pyrophosphate, potassium diphosphate, tkpp, tetrapotassium diphosphate, tetrapotassium pyrophosphate, diphosphoric acid, tetrapotassium salt, tetrapotassium diphosphorate, unii-b9w4019h5g, pyrophosphoric acid, tetrapotassium salt, tetrapotassium phosphonato phosphate | |
RYCLIXPGLDDLTM-UHFFFAOYSA-J | |
tetrapotassium;phosphonato phosphate | |
23740 | |
98% | |
Glass Bottle |
7320-34-5 | |
Crystalline Powder | |
98% | |
K4O7P2 | |
15, 7783 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
[K+].[K+].[K+].[K+].[O-]P([O-])(=O)OP([O-])([O-])=O | |
330.33 | |
330.34 | |
Extra Pure | |
Potassium diphosphate, Extra Pure |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS Signal Word: Warning
EINECSNumber : 230-785-7
RUO – Research Use Only