Propper Chex-All™ II Instant-Seal Pouches
Self-Sealing Chex-All™ II Instant-Seal Pouches used in steam autoclaves or ethylene oxide sterilizers provide a barrier to maintain sterility after the sterilization process. Pre-printed indicators inside and outside the pouch. Triple-sealed paper/plastic film ensures content security.
$212.57 - $1056.23
Specifications
Autoclavable Autoclavable
Closure Type Self-adhesive
Disposable Single-use
For Use With (Application) Indicators on both inside and outside for steam and ethylene oxide (EO) gas
Material Paper, Plastic
View More Specs
Products 7
Catalog Number Mfr. No. Length (Metric) Width (Metric) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) Price Quantity  
15-234-11
View Documents
Propper
02400800
20.3 cm 7.6 cm
Pack of 250 Each for $212.57
Case of 2 Pack for $425.14
15-234-13
View Documents
Propper
02401200
30.5 cm 17.8 cm
Pack of 250 Each for $396.17
Case of 2 Pack for $792.34
15-234-14
View Documents
Propper
02401400
40.6 cm 20.3 cm
Pack of 125 Each for $407.90
Case of 2 Pack for $815.80
15-234-12
View Documents
Propper
02401000
25.4 cm 12.7 cm
Pack of 250 Each for $445.60
Case of 2 Pack for $891.20
15-234-17
View Documents
Propper
02402800
61 cm 8.9 cm
Case of 500 Each for $909.50
15-234-16
View Documents
Propper
02401800
45.7 cm 30.5 cm
Pack of 125 Each for $512.97
Case of 2 Pack for $1,025.94
15-234-15
View Documents
Propper
02401500
38.1 cm 12.7 cm
Case of 500 Each for $1,056.23
Description
Description
Eliminate need for separate indicator strips
  • Self-Sealing Sterilization Pouches: Chex-All™ II sterilization pouches to contain instruments during steam and ethylene oxide (EO, EtO) sterilization.
  • Chex-All™ pouches feature pre-printed process indicators inside and outside the pouch.
  • The steam or ethylene oxide exposure will not affect the 3/4 in.wide adhesive line, guaranteeing a strong flat seal.
  • Self-Sealing Sterilization Pouches: Chex-All™ II is validated for double pouch packaging.
  • After steam sterilization, the chemical indicator changes from yellow to black.
  • After ethylene oxide sterilization, the chemical indicator changes from red to green.
  • Made of 2.5 mm plastic and 73-74 gram paper.
  • Triple sealed paper/plastic film provides excellent sterility protection.
  • Validated for double pouch packaging.
Specifications
Specifications
Autoclavable
Single-use
Paper, Plastic
Self-adhesive
Indicators on both inside and outside for steam and ethylene oxide (EO) gas
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'