Corning™ PYREX™ Low Actinic Class A Volumetric Flask with Glass Standard Taper Stopper
Calibrated “To Contain”
$156.48 - $1990.89
Specifications
Closure Material | Glass |
---|---|
Neck Style | Long |
Graduated | Yes |
Class | A |
Bottom Shape | Flat |
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Closure Type | Tolerance | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Closure Size | Closure Type | Tolerance | Price | Quantity | ||
11-850-22
|
Corning™
55640250FO |
250 mL | 16 | Without Closure | ±0.12 mL |
Case of 12 Each for $1,990.89
|
|
||
11-850-21
|
Corning™
55640200FO |
200 mL | 13 | Without Closure | ±0.1 mL |
Case of 12 Each for $1,724.06
|
|
||
10-227A
|
Corning™
5564025 |
25 mL | 9 | Hollow | ±0.03 mL |
Pack of 6 Each for $787.07
Case of 2 Pack for $1,574.14
|
|
||
10-227B
|
Corning™
5564050 |
50 mL | 9 | Hollow | ±0.05 mL |
Pack of 6 Each for $894.38
Case of 2 Pack for $1,788.76
|
|
||
10-227C
|
Corning™
55640100 |
100 mL | 13 | Hollow | ±0.08 mL |
Each for $156.48
Case of 12 Each for $1,877.76
|
|
||
10-227D
|
Corning™
55640250 |
250 mL | 16 | Hollow | ±0.12 mL |
Pack of 6 Each for $175.50
Case of 2 Pack for $351.00
|
|
||
10-227E
|
Corning™
55640500 |
500 mL | 19 | Hollow | ±0.2 mL |
Case for $371.08
|
|
||
10-227G
|
Corning™
5564010 |
10 mL | 9 | Hollow | ±0.02 mL |
Case of 12 Each for $1,419.62
|
|
||
10-227F
|
Corning™
556401L |
1 L | 22 | Hollow | ±0.3 mL |
Case for $459.03
|
|
||
Description
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of the flask, which retains the mechanical strength, chemical stability, and thermal resistance of PYREX brand labware.
Specifications
Glass | |
Yes | |
Flat |
Long | |
A |