Corning™ PYREX™ VISTA™ Reusable Test Tubes with Beaded Rim
Functionality, economy, and quality
$10.77 - $345.98
Specifications
Design Type | Beaded Rim |
---|---|
Material | Borosilicate Glass |
Sterility | Non-sterile |
Graduated | No |
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Diameter (Metric) | Length (Metric) | Price | Quantity | |||
07-250-112
|
Corning™
7080010 |
3 mL | 10 mm | 75 mm |
Pack of 50 Each for $10.77
Case of 4 Pack for $43.08
|
|
|||
07-250-114
|
Corning™
7080013 |
9 mL | 13 mm | 100 mm |
Pack of 50 Each for $55.09
Case of 4 Pack for $220.36
|
|
|||
07-250-113
|
Corning™
7080012 |
5 mL | 12 mm | 75 mm |
Pack of 50 Each for $79.51
Case of 4 Pack for $318.04
|
|
|||
07-250-116
|
Corning™
7080016 |
20 mL | 16 mm | 150 mm |
Pack of 50 Each for $75.74
Case of 4 Pack for $302.96
|
|
|||
07-250-118
|
Corning™
7080020 |
34 mL | 20 mm | 150 mm |
Pack of 50 Each for $100.49
Case of 4 Pack for $401.96
|
|
|||
07-250-117
|
Corning™
7080018 |
27 mL | 18 mm | 150 mm |
Case of 200 Each for $345.98
|
|
|||
07-250-119
|
Corning™
7080025 |
55 mL | 25 mm | 150 mm |
Pack of 50 Each for $163.79
Case of 4 Pack for $655.16
|
|
|||
07-250-120
|
Corning™
7080025X |
70 mL | 25 mm | 200 mm |
Pack of 50 Each for $187.13
Case of 4 Pack for $748.52
|
|
|||
Description
Functionality, economy, and quality
These test tubes are annealed, resistant to heat and chemically stable. Rims are fire-polished. The 25 mm diameter tubes do not have a marking spot.
Specifications
Beaded Rim | |
Borosilicate Glass | |
Non-sterile | |
No |