missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridoxine hydrochloride, 98%, MP Biomedicals™
$66.69 - $266.68
Chemical Identifiers
CAS | 58-56-0 |
---|---|
Molecular Formula | C8H12ClNO3 |
Molecular Weight (g/mol) | 205.638 |
MDL Number | MFCD00012807 |
InChI Key | ZUFQODAHGAHPFQ-UHFFFAOYSA-N |
Synonym | pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion |
PubChem CID | 6019 |
ChEBI | CHEBI:30961 |
IUPAC Name | 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride |
SMILES | CC1=NC=C(C(=C1O)CO)CO.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10277710
|
MP Biomedicals Inc
0210277710 |
10 g |
N/A
|
|
|||||
ICN10277750
|
MP Biomedicals Inc
0210277750 |
50 g |
N/A
|
|
|||||
ICN10277780
|
MP Biomedicals Inc
0210277780 |
100 g |
N/A
|
|
|||||
Chemical Identifiers
58-56-0 | |
205.638 | |
ZUFQODAHGAHPFQ-UHFFFAOYSA-N | |
6019 | |
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride |
C8H12ClNO3 | |
MFCD00012807 | |
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion | |
CHEBI:30961 | |
CC1=NC=C(C(=C1O)CO)CO.Cl |
Specifications
58-56-0 | |
1.43g/mL | |
C8H12ClNO3 | |
10 g | |
ZUFQODAHGAHPFQ-UHFFFAOYSA-N | |
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride | |
6019 | |
205.64 | |
Powder |
White | |
>98% | |
MFCD00012807 | |
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion | |
CC1=NC=C(C(=C1O)CO)CO.Cl | |
205.638 | |
CHEBI:30961 | |
0.98 | |
Pyridoxine hydrochloride |
Safety and Handling
EINECSNumber : 200-386-2
Recommended Storage : Store at Room Temperature.