Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap
Provides additional protection for light-sensitive contents by blocking UV rays. Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap is useful for wide range of applications including pharmaceutical, chemical and environmental.
$138.82 - $2451.00
Specifications
Cleaning Procedure Vacuum and ionized
Color Amber
Material Glass
Closure Color Green
Closure Material Thermoset
View More Specs
Products 13
Catalog Number Mfr. No. Capacity (Metric) Closure Size Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Closure Size Packaging Quantity Price Quantity  
02-991-851
View Documents
Qorpak™
GLC11310
15 mL 28-400 mm Conventional pack 48/Cs.
Case of 48 Each for $328.12
36-103-0212
View Documents
Qorpak™
GLC02080
30 mL 28-400 mm Bulk pack 432/Cs.
Case of 432 Each for $2,451.00
36-104-4715
View Documents
Qorpak™
GLC02083
30 mL 28-400 mm Conventional pack 24/Cs.
Case of 24 Each for $138.82
02992095
View Documents
Qorpak™
GLC02098
60 mL 33-400 mm Conventional pack 24/Cs.
Case of 24 Each for $174.15
36-103-0213
View Documents
Qorpak™
GLC02189
120 mL 38-400 mm Bulk pack 180/Cs.
Case of 180 Each for $1,463.81
02-992-093
View Documents
Qorpak™
GLC02192
120 mL 38-400 mm Conventional pack 24/Cs.
Case of 24 Each for $201.88
GLC02116
View Documents
Qorpak™
GLC02116
250 mL 45-400 mm Bulk pack 84/Cs.
Case of 84 Each for $848.03
36-104-4716
View Documents
Qorpak™
GLC02120
250 mL 45-400 mm Conventional pack 24/Cs.
Case of 24 Each for $255.56
02-992-098
View Documents
Qorpak™
GLC02137
500 mL 53-400 mm Conventional pack 12/Cs.
Case of 12 Each for $169.44
GLC02133
View Documents
Qorpak™
GLC02133
500 mL 53-400 mm Bulk pack 60/Cs.
Case of 60 Each for $786.63
02-992-565
View Documents
Qorpak™
GLC02164
950 mL 53-400 mm Conventional pack 12/Cs.
Case of 12 Each for $204.29
GLC07007
View Documents
Qorpak™
GLC07007
1250 mL 70-400 mm Conventional pack 6/Cs.
Case of 6 Each for $199.41
36-104-4717
View Documents
Qorpak™
GLC07354
2500 mL 70-400 mm Conventional pack 12/Cs.
Case of 12 Each for $674.85
Description
Description
Provides additional protection for light-sensitive contents by blocking UV rays. Qorpak™ Amber Wide Mouth Packer Bottles, Vacuum and Ionized with Thermoset F217 and PTFE Cap is useful for wide range of applications including pharmaceutical, chemical and environmental.
  • Air inside the bottle is odor-free, tasteless and does not react with product
  • Separate washing not required, thereby saving time
  • Final finished product kept clean and dust-free
  • Contaminants such as loose dirt, carton lint, aerosols, and fine glass particles are removed by the Vacuum and Ionization process
  • Bottles are made of Type III, soda lime glass.
Specifications
Specifications
Vacuum and ionized
Glass
Thermoset
Amber
Green
Screw Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'