missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 4-aminohippurate hydrate, 98%, Thermo Scientific Chemicals

$94.71 - $1,098.07

Chemical Identifiers

CAS 206658-83-5
Molecular Formula C9H10N2NaO3+
Molecular Weight (g/mol) 217.18
MDL Number MFCD00150723
InChI Key UNZMYCAEMNVPHX-UHFFFAOYSA-N
Synonym aminohippurate sodium
PubChem CID 57465078
IUPAC Name sodium;2-[(4-aminobenzoyl)amino]acetic acid
SMILES C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1577506
View Documents
Thermo Scientific Chemicals
A1577506
5 g N/A
 
AAA1577514
View Documents
Thermo Scientific Chemicals
A1577514
25 g N/A
 
AAA1577522
View Documents
Thermo Scientific Chemicals
A1577522
100 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

206658-83-5
217.18
UNZMYCAEMNVPHX-UHFFFAOYSA-N
57465078
C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]
C9H10N2NaO3+
MFCD00150723
aminohippurate sodium
sodium;2-[(4-aminobenzoyl)amino]acetic acid

Specifications

206658-83-5
C9H10N2NaO3+
5 g
UNZMYCAEMNVPHX-UHFFFAOYSA-N
sodium;2-[(4-aminobenzoyl)amino]acetic acid
57465078
98%
123°C to 127°C
MFCD00150723
aminohippurate sodium
C1=CC(=CC=C1C(=O)NCC(=O)O)N.[Na+]
217.18
216.17 (Anhydrous)
Sodium 4-aminohippurate hydrate
Safety and Handling

Safety and Handling

EINECSNumber : 202-309-8

RTECSNumber : MR8157200

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only