missing translation for 'onlineSavingsMsg'
Learn More

Sodium dihydrogen phosphate dihydrate, 98+%

CAS: 13472-35-0 | H6NaO6P | 156.006 g/mol

$62.14 - $199.48

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
MDL Number MFCD00149209
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
Synonym sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
PubChem CID 23673460
IUPAC Name sodium;dihydrogen phosphate;dihydrate
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1131622
View Documents
Thermo Scientific Chemicals
A1131622
100 g
Each for $62.14
Only null left
 
AAA1131636
View Documents
Thermo Scientific Chemicals
A1131636
500 g
Each for $86.14
Only null left
 
AAA113160E
View Documents
Thermo Scientific Chemicals
A113160E
2500 g
Each for $199.48
Only null left
 
Description

Description

Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium dihydrogen phosphate dihydrate acts as buffering capacity reagent in molecular biology, biochemistry and chromatography. It is used in the preparation of biological buffers. It is also used in the purification of antibodies, as a laxative and, in combination with other sodium phosphates

Solubility
Soluble in water.(1 M at 20°C)

Notes
Hygroscopic. Store under inert gas. Incompatible with oxidizing agents and water.
Specifications

Chemical Identifiers

13472-35-0
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
sodium;dihydrogen phosphate;dihydrate
H6NaO6P
VBJGJHBYWREJQD-UHFFFAOYSA-M
23673460

Specifications

13472-35-0
H6NaO6P
MFCD00149209
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh
VBJGJHBYWREJQD-UHFFFAOYSA-M
sodium;dihydrogen phosphate;dihydrate
23673460
99%
1.91
100 g
NaH2PO4·2H2O
14,8660
Soluble in water.(1 M at 20°C)
O.O.OP(=O)(O)[O-].[Na+]
156.006
156.01 (119.98 Anhydrous)
Hygroscopic
Sodium dihydrogen phosphate dihydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-449-2

RTECSNumber : WA1900000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only