Eisco™ Solid ASTM Neoprene Stoppers
Pack of ten solid ASTM neoprene stoppers. Made from long lasting neoprene rubber. Oil resistant, use up to 250°F / 120°C. Suitable for use with petroleum products, minerals, oils, fats as well as most inorganic acids.
$7.71 - $1292.65
Specifications
Certifications/Compliance | ASTM |
---|---|
Description | Pack of 10 |
Autoclavable | Autoclavable |
Color | Blue |
Material | Neoprene |
Catalog Number | Mfr. No. | Diameter (Metric) | Diameter (Metric) Bottom | Height (Metric) | Stopper Number | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Diameter (Metric) Bottom | Height (Metric) | Stopper Number | Price | Quantity | ||
S41055
|
Eisco™
CH0321A |
12.7 mm | 8.2 mm | 25 mm | #000 |
Pack of 10 Each for $7.71
|
|
||
S27040
|
Eisco™
CH0321Q |
50 mm | 42 mm | 25 mm | #10 |
Each for $166.26
|
|
||
S41057
|
Eisco™
CH0321B |
15 mm | 10 mm | 25 mm | #00 |
Pack of 10 Each for $12.21
|
|
||
S41059
|
Eisco™
CH0321C |
17 mm | 13 mm | 25 mm | #0 |
Pack of 10 Each for $17.47
|
|
||
S41062
|
Eisco™
CH0321D |
19 mm | 14 mm | 25 mm | #1 |
Pack of 10 Each for $19.24
|
|
||
S41065
|
Eisco™
CH0321E |
20 mm | 16 mm | 25 mm | #2 |
Pack of 10 Each for $19.82
|
|
||
S41069
|
Eisco™
CH0321F |
24 mm | 18 mm | 25 mm | #3 |
Pack of 10 Each for $27.12
|
|
||
S41072
|
Eisco™
CH0321G |
26 mm | 20 mm | 25 mm | #4 |
Pack of 10 Each for $32.47
|
|
||
S41075
|
Eisco™
CH0321H |
27 mm | 23 mm | 25 mm | #5 |
Pack of 10 Each for $40.44
|
|
||
S41078
|
Eisco™
CH0321I |
28 mm | 24 mm | 25 mm | #5.5 |
Pack of 10 Each for $46.09
|
|
||
S41081
|
Eisco™
CH0321J |
32 mm | 26 mm | 25 mm | #6 |
Pack of 10 Each for $52.24
|
|
||
S41084
|
Eisco™
CH0321K |
34 mm | 27 mm | 25 mm | #6.5 |
Pack of 10 Each for $59.82
|
|
||
S41087
|
Eisco™
CH0321L |
37 mm | 30 mm | 25 mm | #7 |
Pack of 10 Each for $70.41
|
|
||
S41090
|
Eisco™
CH0321M |
39 mm | 31 mm | 25 mm | #7.5 |
Pack of 10 Each for $83.26
|
|
||
S41093
|
Eisco™
CH0321N |
41 mm | 33 mm | 25 mm | #8 |
Pack of 10 Each for $86.56
|
|
||
S41095
|
Eisco™
CH0321O |
43 mm | 36 mm | 25 mm | #8.5 |
Pack of 10 Each for $97.32
|
|
||
S41098
|
Eisco™
CH0321P |
45 mm | 37 mm | 25 mm | #9 |
Pack of 10 Each for $108.06
|
|
||
S41103
|
Eisco™
CH0321R |
53 mm | 45 mm | 25 mm | #10.5 |
Pack of 10 Each for $145.88
|
|
||
S41106
|
Eisco™
CH0321S |
56 mm | 48 mm | 25 mm | #11 |
Pack of 10 Each for $171.26
|
|
||
S41109
|
Eisco™
CH0321T |
63 mm | 50 mm | 25 mm | #11.5 |
Pack of 10 Each for $201.82
|
|
||
S41112
|
Eisco™
CH0321U |
64 mm | 54 mm | 25 mm | #12 |
Pack of 10 Each for $217.97
|
|
||
S41116
|
Eisco™
CH0321V |
68 mm | 58 mm | 25 mm | #13 |
Pack of 10 Each for $253.00
|
|
||
S41119
|
Eisco™
CH0321W |
75 mm | 62 mm | 35 mm | #13.5 |
Pack of 10 Each for $456.56
|
|
||
S41124
|
Eisco™
CH0321X |
90 mm | 75 mm | 39 mm | #14 |
Pack of 10 Each for $533.53
|
|
||
S41127
|
Eisco™
CH0321Y |
103 mm | 83 mm | 39 mm | #15 |
Pack of 10 Each for $814.06
|
|
||
S41130
|
Eisco™
CH0321Z |
127 mm | 90 mm | 50 mm | #16 |
Pack of 10 Each for $1,292.65
|
|
||
Description
Warranty
1 year limited manufacturer warranty
Specifications
ASTM | |
Autoclavable | |
Neoprene | |
Conical | |
Reusable | |
Petroleum products, minerals, oils and fats, as well as most inorganic acids and bases |
Pack of 10 | |
Blue | |
Steri-stopper | |
Non-sterile | |
Stopper for Labware | |
-40 to 130°C |