missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trifluoroacetic Acid, 99.9%, Spectrum™ Chemical
$160.76 - $12387.32
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18614560
|
Spectrum Chemical Mfg Cor
TR11925GM |
25 g | Amber Glass Bottle |
Each for $160.76
|
|
||||
18614558
|
Spectrum Chemical Mfg Cor
TR119100GM |
100 g | Amber Glass Bottle |
Each for $261.76
|
|
||||
18614562
|
Spectrum Chemical Mfg Cor
TR119500GM |
500 g | Amber Glass Bottle |
Each for $709.56
|
|
||||
18614559
|
Spectrum Chemical Mfg Cor
TR1191KG |
1 kg | Amber Glass Bottle |
Each for $1,186.00
|
|
||||
18614563
|
Spectrum Chemical Mfg Cor
TR11925KG |
2.5 kg | Glass Bottle |
Each for $1,902.94
|
|
||||
18614561
|
Spectrum Chemical Mfg Cor
TR11927KGBL |
27 kg | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
76-05-1 |
Specifications
76-05-1 | |
99.9% | |
25 g |
1 | |
Reagent | |
Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.