Hamilton™ 700 Microliter Syringes: N Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
$63.25 - $196.58
Specifications
Autoclavable | Not Autoclavable |
---|---|
For Use With (Application) | For dispensing low volumes to achieve the highest level of accuracy and precision possible |
Length (English) Needle | 2 in. |
Product Type | Microliter Syringe |
Catalog Number | Mfr. No. | Needle Point Style | Diameter (Metric) Inner Needle | Diameter (Metric) Outer Needle | Volume (Metric) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Point Style | Diameter (Metric) Inner Needle | Diameter (Metric) Outer Needle | Volume (Metric) | Price | Quantity | ||
14-815-291
|
Hamilton™
HAM80765 |
Blunt | 0.41 mm | 0.72 mm | 250 μL |
Each for $155.55
|
|
||
14-815-292
|
Hamilton™
HAM80865 |
Blunt | 0.41 mm | 0.72 mm | 500 μL |
Each for $176.08
|
|
||
14-813-86
|
Hamilton™
HAM80839 |
Cone | 0.41 mm | 0.72 mm | 500 μL |
Each for $196.58
|
|
||
14-813-6
|
Hamilton™
HAM80339 |
Cone | 0.13 mm | 0.47 mm | 10 μL |
Each for $83.75
|
|
||
14-813-24
|
Hamilton™
HAM80439 |
Cone | 0.15 mm | 0.72 mm | 25 μL |
Each for $114.53
|
|
||
14-816-208B
|
Hamilton™
HAM80383 |
Blunt | 0.13 mm | 0.72 mm | 10 μL |
Each for $63.25
|
|
||
14-824-32
|
Hamilton™
HAM80565 |
Blunt | 0.15 mm | 0.72 mm | 50 μL |
Each for $116.25
|
|
||
14-813-091
|
Hamilton™
HAM80621 |
Blunt | 0.41 mm | 0.72 mm | 100 μL |
Each for $130.53
|
|
||
14-815-288
|
Hamilton™
HAM80465 |
Blunt | 0.15 mm | 0.72 mm | 25 μL |
Each for $95.72
|
|
||
14-815-287
|
Hamilton™
HAM80365 |
3 | 0.15 mm | 0.72 mm | 10 μL |
Each for $70.09
|
|
||
14-824-5
|
Hamilton™
HAM80500 |
Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm | 50 μL |
Each for $117.94
|
|
||
14-813-090
|
Hamilton™
HAM80384 |
Sharp, Bevel, Curved, Non-coring | 0.13 mm | 0.47 mm | 10 μL |
Each for $66.66
|
|
||
14-824-7
|
Hamilton™
HAM80400 |
Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm | 25 μL |
Each for $95.72
|
|
||
14-824-713
|
Hamilton™
87919 |
3 | 0.13 mm | 0.483 mm | 5 μL |
Each for $138.26
|
|
||
14-813-71
|
Hamilton™
HAM80739 |
Cone | 0.41 mm | 0.72 mm | 250 μL |
Each for $166.09
|
|
||
14-813-55
|
Hamilton™
HAM80639 |
Cone | 0.15 mm | 0.72 mm | 100 μL |
Each for $155.55
|
|
||
14-824-6
|
Hamilton™
HAM80600 |
Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm | 100 μL |
Each for $136.75
|
|
||
14-816213B
|
Hamilton™
HAM80521 |
Blunt | 0.41 mm | 0.72 mm | 50 μL |
Each for $104.83
|
|
||
14-815-290
|
Hamilton™
HAM80665 |
Blunt | 0.15 mm | 0.72 mm | 100 μL |
Each for $136.75
|
|
||
14-824-2
|
Hamilton™
HAM80700 |
Sharp, Bevel, Curved, Non-coring | 0.15 mm | 0.72 mm | 250 μL |
Each for $162.40
|
|
||
14-824-9
|
Hamilton™
HAM80800 |
Sharp, Bevel, Curved, Non-coring | 0.41 mm | 0.72 mm | 500 μL |
Each for $177.77
|
|
||
14-813-40
|
Hamilton™
HAM80539 |
Cone | 0.15 mm | 0.72 mm | 50 μL |
Each for $136.75
|
|
||
Description
- Original hand-fitted Hamilton syringe
- Ideal for dispensing volumes from 0.5 µL up to 500 µL
- A variety of different terminations and needle options are available in this series
Specifications
Not Autoclavable | |
For dispensing low volumes to achieve the highest level of accuracy and precision possible | |
2 in. | |
Microliter Syringe |