missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, 99%

Di-n-butylamine, 99%, C8H19N, CAS Number-111-92-2, di-normal-butylamine, n-dibutylamine, n-butyl-1-butanamine, di-n-butyl amine, dibutilamina, dibutylamine, dibutyl amine, di-n-butylamine, dibutyl-amine, 1-butanamine, n-butyl, 2.5L, 98.5% min. (GC), 04, 157, 159.0 deg.C, Undesignated, 0.7600g/mL | CAS: 111-92-2 | C8H19N | 129.24 g/mol

$109.04 - $315.36

Chemical Identifiers

CAS 111-92-2
MDL Number MFCD00009429
InChI Key JQVDAXLFBXTEQA-UHFFFAOYSA-N
Synonym dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
PubChem CID 8148
IUPAC Name N-butylbutan-1-amine
SMILES CCCCNCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC112950500
SDS View Documents
Thermo Scientific Chemicals
112950500
50 mL Glass bottle
Each for $109.04
Only null left
 
AC112950010
SDS View Documents
Thermo Scientific Chemicals
112950010
1 L Glass Bottle
Each for $165.21
Only null left
 
AC112950025
SDS View Documents
Thermo Scientific Chemicals
112950025
2.5 L Glass bottle
Each for $315.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-92-2
JQVDAXLFBXTEQA-UHFFFAOYSA-N
8148
CCCCNCCCC
MFCD00009429
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
N-butylbutan-1-amine

Specifications

111-92-2
0.7600g/mL
39°C
98.5% min. (GC)
C8H19N
[CH3(CH2)3]2NH
50 mL
01,209
15, 3037
Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
CCCCNCCCC
129.24
0.9 mPa.s (20°C)
99%
Di-n-butylamine
-62.0°C
159.0°C
Authentic
Glass bottle
1.4160 to 1.4180
MFCD00009429
04, 157
0.76
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
JQVDAXLFBXTEQA-UHFFFAOYSA-N
N-butylbutan-1-amine
8148
129.24
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fre

GHS Signal Word: Danger

EINECSNumber : 203-921-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only