Learn More
Fisherbrand™ General Purpose Liquid-in-Glass Partial Immersion Thermometers
Fisherbrand™ General Purpose Liquid-in-Glass Partial Immersion Thermometers are ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories and the pharmaceutical industry.
$19.88 - $54.50
Specifications
Certifications/Compliance ISO/IEC 17025:2017, A2LA, NIST
For Use With (Application) Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Immersion Type Partial Immersion
Material Glass
Thermometer Fill Blue Organic Liquid
View More Specs
Products 11
Catalog Number Mfr. No. Temperature Range (Metric) Temperature Range (English) Length (Metric) Overall Price Quantity  
Catalog Number Mfr. No. Temperature Range (Metric) Temperature Range (English) Length (Metric) Overall Price Quantity  
13201521
View Documents
Fisherbrand™
B607601220
-100°C to +50°C n.a. 305 mm N/A
Only null left
13201555 Edge
Accessories View Documents
Fisherbrand™
B607000920
-10°C to +260°C n.a. 355 mm N/A
Only null left
Edge
13201640
View Documents
Fisherbrand™
B607000120
-10°C to +110°C n.a. 200 mm N/A
Only null left
13201643
View Documents
Fisherbrand™
B607000320
-35°C to +50°C n.a. 300 mm N/A
Only null left
13201644
View Documents
Fisherbrand™
B607000420
-20°C to +110°C n.a. 300 mm N/A
Only null left
13201641
View Documents
Fisherbrand™
B607000220
-10°C to +150°C n.a. 200 mm N/A
Only null left
13201645
View Documents
Fisherbrand™
B607000520
-20°C to +150°C n.a. 300 mm N/A
Only null left
13-201-646
View Documents
Fisherbrand™
B607000620
n.a. -40°F to +120°F 300 mm N/A
Only null left
13-201-647
View Documents
Fisherbrand™
B607000720
n.a. 0°F to 230°F 300 mm N/A
Only null left
13-201-649
View Documents
Fisherbrand™
B607001020
n.a. 20°F to 500°F 355 mm N/A
Only null left
13201648
View Documents
Fisherbrand™
B607000820
n.a. 0°F to 300°F 300 mm N/A
Only null left
Description
Description
  • Traceable to NIST, individually serialized
  • Feature blue organic liquid fill against lead-free white back glass
  • Include multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability
  • Tested and inspected by our USA quality team
Specifications
Specifications
ISO/IEC 17025:2017, A2LA, NIST
Partial Immersion
Blue Organic Liquid
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Glass
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'