Fisherbrand™ Polymethylpentene Graduated Cylinders
Clear, shatterproof polymethylpentene polymer.
$7.67 - $60.26
Specifications
Material PMP, PMP
Class B, B
Autoclavable Autoclavable, Autoclavable
Max. Temperature 135°C, 135°C
Base Type Hexagonal, Hexagonal
View More Specs
Products 6
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity  
S32812
View Documents
Fisherbrand™
237755EMD
50 mL, 50 mL 1 mL, 1 mL
Each for $12.64
Case of 18 Each for $227.52
S32810
View Documents
Fisherbrand™
237735EMD
10 mL, 10 mL 0.2 mL, 0.2 mL
Each for $7.67
Case of 24 Each for $184.08
S32813
View Documents
Fisherbrand™
237765EMD
100 mL, 100 mL 1 mL, 1 mL
Each for $17.15
Case of 12 Each for $205.80
S32816
View Documents
Fisherbrand™
237895EMD
1000 mL, 1000 mL 10 mL, 10 mL
Each for $60.26
Case of 6 Each for $361.56
S32811
View Documents
Fisherbrand™
237745EMD
25 mL, 25 mL 0.5 mL, 0.5 mL
Each for $10.72
Case of 18 Each for $192.96
S32814
View Documents
Fisherbrand™
237775EMD
250 mL, 250 mL 2 mL, 2 mL
Each for $24.18
Case of 12 Each for $290.16
Description
Description
  • Withstand autoclaving temperatures of up to 175°C
  • Hexagonal bases improve stability
Specifications
Specifications
PMP, PMP
Autoclavable, Autoclavable
Hexagonal, Hexagonal
B, B
135°C, 135°C
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'