missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Remco™ Polypropylene Tools

Tough, nonsparking polypropylene, highly resistant to chemical attack

$9.42 - $11.26

Specifications

Certifications/Compliance FDA
Color White
For Use With (Application) Provide scratch-free use on metal surfaces, ideal for cleanup of explosive materials and volatile liquids
Material Polypropylene
Temperature Range (English) 176°F
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Size Price Quantity  
Catalog Number Mfr. No. Product Type Size Price Quantity  
17-931A
View Documents
Remco™
64005
Scoop Small
Each for $9.42
Pack of 12 Each for $104.76
 
17-931B
View Documents
Remco™
6500
Scoop Large
Each for $11.26
Pack of 12 Each for $125.16
 
29-635-580
View Documents
Remco™
69615
Hand Scraper Small
Each for $9.45
 
17-932-4
View Documents
Remco™
69625
Hand Scraper Large
Each for $10.71
 
Description

Description

  • Tough, nonsparking polypropylene tools are highly resistant to chemical attack and corrosion—ideal for cleanup of explosive materials and volatile liquids
  • Resilient and lightweight for easy handling
  • Withstand temperatures up to 80°C (176°F)
  • Economical enough for disposal after single use, or decontaminate and use again
  • Pan-type and two-piece green shovels

Specifications

Specifications

FDA
Provide scratch-free use on metal surfaces, ideal for cleanup of explosive materials and volatile liquids
176°F
White
Polypropylene
80°C
SDS
missing translation for 'documents'

missing translation for 'documents'